siRNA Details
Gene Symbol: MESDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022566.2 | NP_072088.1 | 1 | 2340 | ||
GenBank mRNA | AY007810.1 | - | 1 | 1662 | ||
BC084554.1 | - | 1 | 1199 | 6088754 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
