siRNA Details
Gene Symbol: GTF2H2C_2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042490.3 | NP_001035955.1 | 3, 4 | 352 | ||
GenBank mRNA | BC064557.1 | - | 2, 3 | 232 | 5549265 | |
BC065021.1 | - | 3, 4 | 353 | 6047020 | ||
BX647445.1 | - | 2, 3 | 117 |
Gene Symbol: GTF2H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001515.3 | NP_001506.1 | 2, 3 | 274 | ||
XM_017009403.1 | XP_016864892.1 | 3, 4 | 883 | |||
XM_017009404.1 | XP_016864893.1 | 3, 4 | 371 | |||
XM_017009405.1 | XP_016864894.1 | 3, 4 | 226 | |||
XM_017009406.1 | XP_016864895.1 | 2, 3 | 129 | |||
GenBank mRNA | AF078847.1 | - | 2, 3 | 259 | ||
AK296578.1 | - | 2, 3 | 278 | |||
BC005345.1 | - | 2, 3 | 132 | |||
BM083743.1 | - | 2, 3 | 274 | |||
BT006773.1 | - | 1, 2 | 64 | |||
Z30094.1 | - | 1, 2 | 64 |
Gene Symbol: GTF2H2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033417.1 | 2, 3 | 278 | |||
GenBank mRNA | BC171919.1 | - | 2, 3 | 207 | 9054573 | |
BX537982.1 | - | 2, 3 | 118 |
Gene Symbol: GTF2H2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098728.1 | NP_001092198.1 | 2, 3 | 277 | ||
XM_005248588.4 | XP_005248645.1 | 2, 3 | 204 | |||
XM_005248590.3 | XP_005248647.1 | 2, 3 | 142 | |||
XM_006714688.3 | XP_006714751.1 | 2, 3 | 195 | |||
XM_011543615.1 | XP_011541917.1 | 2, 3 | 277 | |||
XM_017009804.1 | XP_016865293.1 | 1, 2 | 97 | |||
GenBank mRNA | AK292361.1 | - | 2, 3 | 256 | ||
AK315282.1 | - | 2, 3 | 277 | |||
BC105980.1 | - | 2, 3 | 135 | 4445643 | ||
BC157894.1 | - | 2, 3 | 207 | |||
BC171860.1 | - | 3, 4 | 269 | 9054514 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
