siRNA Details
Gene Symbol: GTF2H2C_2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042490.3 | NP_001035955.1 | 17 | 1710 | ||
GenBank mRNA | BC064557.1 | - | 16 | 1590 | 5549265 | |
BC065021.1 | - | 17 | 1711 | 6047020 |
Gene Symbol: GTF2H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001515.3 | NP_001506.1 | 16 | 1632 | ||
XM_017009403.1 | XP_016864892.1 | 17 | 2241 | |||
XM_017009404.1 | XP_016864893.1 | 17 | 1729 | |||
XM_017009405.1 | XP_016864894.1 | 17 | 1584 | |||
XM_017009406.1 | XP_016864895.1 | 16 | 1487 | |||
GenBank mRNA | AF078847.1 | - | 16 | 1617 |
Gene Symbol: GTF2H2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033417.1 | 15 | 1599 | |||
GenBank mRNA | BC171919.1 | - | 15 | 1528 | 9054573 |
Gene Symbol: GTF2H2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098728.1 | NP_001092198.1 | 16 | 1635 | ||
XM_005248588.4 | XP_005248645.1 | 16 | 1562 | |||
XM_006714688.3 | XP_006714751.1 | 16 | 1553 | |||
XM_011543615.1 | XP_011541917.1 | 15 | 1595 | |||
XM_017009804.1 | XP_016865293.1 | 14 | 1418 | |||
GenBank mRNA | AK292361.1 | - | 16 | 1614 | ||
BC157894.1 | - | 16 | 1565 | |||
BC171860.1 | - | 17 | 1627 | 9054514 | ||
DQ786295.1 | - | 12 | 1255 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
