siRNA Details
Gene Symbol: KRTAP21-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181619.1 | NP_853650.1 | 1 | 83 | ||
GenBank mRNA | AB096960.1 | - | 1 | 112 | ||
BC105029.1 | - | 1 | 133 | 8144032 | ||
BC105031.1 | - | 1 | 133 | 8144034 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
