siRNA Details
Gene Symbol: PPAN-P2RY11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040664.2 | NP_001035754.1 | 3 | 453 | ||
NM_001198690.1 | NP_001185619.1 | 3 | 453 | |||
GenBank mRNA | AJ300588.1 | - | 3 | 281 | ||
BC171852.1 | - | 3 | 453 |
Gene Symbol: PPAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020230.5 | NP_064615.3 | 3 | 453 | ||
GenBank mRNA | AJ292529.1 | - | 3 | 308 | ||
BC009833.2 | - | 3 | 325 | 3957286 | ||
BC033202.1 | - | 3 | 387 | 4871446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
