siRNA Details
Gene Symbol: PRR5-ARHGAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181334.5 | NP_851851.3 | 15 | 1668 | ||
GenBank mRNA | AF195968.1 | - | 14 | 1259 | ||
AL355192.1 | - | 17 | 1628 | |||
BC132755.1 | - | 13 | 1159 | 40146777 | ||
BC132757.1 | - | 13 | 1159 | 40146779 | ||
BC144382.1 | - | 13 | 1210 | 9052903 |
Gene Symbol: ARHGAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017526.1 | NP_001017526.1 | 13 | 1232 | ||
NM_001198726.1 | NP_001185655.1 | 11 | 1035 | |||
NM_181335.2 | NP_851852.2 | 12 | 1139 | |||
GenBank mRNA | AF544240.1 | - | 11 | 998 | ||
AK001306.1 | - | 13 | 1232 | |||
BC010490.2 | - | 10 | 983 | |||
BC048280.1 | - | 12 | 1091 | 5190702 | ||
BC093675.1 | - | 2 | 77 | |||
BC111953.1 | - | 2 | 77 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
