siRNA Details
Gene Symbol: TRIM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003827.1 | NP_001003827.1 | 8 | 2205 | ||
NM_021616.5 | NP_067629.2 | 8 | 2209 | |||
GenBank mRNA | AB039902.1 | - | 8 | 2213 | ||
AF220143.1 | - | 8 | 2192 | |||
AF220144.1 | - | 9 | 2125 | |||
AK027876.1 | - | 8 | 2181 | |||
AL583914.1 | - | 7 | 1814 |
Gene Symbol: TRIM6-TRIM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003819.3 | NP_001003819.1 | 14 | 3373 | ||
GenBank mRNA | AB039903.1 | - | 14 | 3281 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
