siRNA Details
Gene Symbol: SMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000344.3 | NP_000335.1 | 7 | 956 | ||
NM_001297715.1 | NP_001284644.1 | 7 | 956 | |||
NM_022874.2 | NP_075012.1 | 6 | 860 | |||
XM_011543596.1 | XP_011541898.1 | 7 | 835 | |||
XM_011543597.1 | XP_011541899.1 | 6 | 640 | |||
XM_011543598.2 | XP_011541900.1 | 5 | 555 | |||
XM_017009786.1 | XP_016865275.1 | 6 | 739 | |||
GenBank mRNA | AK298203.1 | - | 6 | 640 | ||
BC062723.1 | - | 7 | 812 | 4250429 | ||
JQ657798.1 | - | 4 | 449 | |||
JQ657799.1 | - | 4 | 449 | |||
JQ657800.1 | - | 4 | 344 | |||
JQ657802.1 | - | 4 | 449 | |||
JQ657803.1 | - | 3 | 248 | |||
JQ690867.1 | - | 4 | 344 | |||
JQ690868.1 | - | 3 | 248 | |||
JQ745297.1 | - | 3 | 191 | |||
U18423.1 | - | 7 | 826 |
Gene Symbol: SMN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017411.3 | NP_059107.1 | 7 | 956 | ||
NM_022875.2 | NP_075013.1 | 7 | 956 | |||
NM_022876.2 | NP_075014.1 | 6 | 860 | |||
NM_022877.2 | NP_075015.1 | 6 | 860 | |||
XM_011543599.1 | XP_011541901.1 | 7 | 835 | |||
XM_011543600.2 | XP_011541902.1 | 6 | 640 | |||
XM_011543601.1 | XP_011541903.1 | 6 | 640 | |||
XM_011543602.2 | XP_011541904.1 | 5 | 556 | |||
XM_011543603.2 | XP_011541905.1 | 5 | 556 | |||
XM_017009787.1 | XP_016865276.1 | 7 | 835 | |||
GenBank mRNA | AK289669.1 | - | 7 | 839 | ||
BC000908.2 | - | 7 | 825 | 2985046 | ||
BC015308.1 | - | 7 | 823 | 4427000 | ||
BC070242.1 | - | 7 | 802 | 4673318 | ||
CN272420.1 | - | 5 | 659 | |||
JQ690861.1 | - | 4 | 344 | |||
JQ690862.1 | - | 4 | 344 | |||
JQ690863.1 | - | 3 | 248 | |||
JQ690864.1 | - | 4 | 392 | |||
JQ690865.1 | - | 4 | 449 | |||
JQ690866.1 | - | 3 | 248 | |||
U21914.1 | - | 7 | 790 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
