siRNA Details
Gene Symbol: SMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000344.3 | NP_000335.1 | 8 | 1011 | ||
NM_022874.2 | NP_075012.1 | 7 | 915 | |||
XM_011543597.1 | XP_011541899.1 | 7 | 695 | |||
XM_011543598.2 | XP_011541900.1 | 6 | 610 | |||
GenBank mRNA | AK298203.1 | - | 7 | 695 | ||
BC062723.1 | - | 8 | 867 | 4250429 | ||
JQ657798.1 | - | 5 | 504 | |||
JQ657800.1 | - | 5 | 399 | |||
JQ657802.1 | - | 5 | 504 | |||
JQ657803.1 | - | 4 | 303 | |||
JQ732166.1 | - | 4 | 393 | |||
JQ745297.1 | - | 4 | 246 | |||
U18423.1 | - | 8 | 881 |
Gene Symbol: SMN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CN272420.1 | - | 6 | 714 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
