siRNA Details
Gene Symbol: C12orf73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135570.1 | NP_001129042.1 | 3 | 324 | ||
XM_011538718.2 | XP_011537020.1 | 4 | 1179 | |||
XM_017019916.1 | XP_016875405.1 | 3 | 782 | |||
XM_017019917.1 | XP_016875406.1 | 3 | 387 | |||
GenBank mRNA | AA563681.1 | - | 3 | 252 | ||
AK024037.1 | - | 2 | 619 | |||
AL359596.1 | - | 2 | 347 | |||
BC039467.1 | - | 3 | 252 | |||
BC044618.2 | - | 3 | 360 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
