siRNA Details
Gene Symbol: UBE2V1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032288.2 | NP_001027459.1 | 4 | 2038 | ||
NM_001257393.1 | NP_001244322.1 | 5 | 2213 | |||
NM_001257394.1 | NP_001244323.1 | 3 | 2272 | |||
NM_001257395.1 | NP_001244324.1 | 4 | 2029 | |||
NM_001257396.1 | NP_001244325.1 | 3 | 1912 | |||
NM_001257397.1 | NP_001244326.1 | 5 | 2121 | |||
NM_001257398.1 | NP_001244327.1 | 5 | 2177 | |||
NM_001257399.1 | NP_001244328.1 | 5 | 2156 | |||
NM_001282575.2 | NP_001269504.1 | 5 | 2473 | |||
NM_001282576.1 | NP_001269505.1 | 5 | 2312 | |||
NM_001282577.2 | NP_001269506.1 | 5 | 2090 | |||
NM_001282578.2 | NP_001269507.1 | 4 | 2227 | |||
NM_001282579.1 | NP_001269508.1 | 5 | 2138 | |||
NM_021988.5 | NP_068823.2 | 4 | 2421 | |||
NM_022442.5 | NP_071887.1 | 4 | 2064 | |||
NM_199144.2 | NP_954595.1 | 5 | 2344 | |||
NR_047554.1 | 3 | 1889 | ||||
GenBank mRNA | AL110132.1 | - | 2 | 4054 | ||
BX647511.1 | - | 3 | 1857 | |||
BX648147.1 | - | 3 | 1865 | |||
U97279.1 | - | 1 | 2305 |
Gene Symbol: TMEM189-UBE2V1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199203.2 | NP_954673.1 | 8 | 2798 | ||
GenBank mRNA | CR738799.1 | - | 1 | 346 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
