siRNA Details
Gene Symbol: UBE2V1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032288.2 | NP_001027459.1 | 4 | 1344 | ||
NM_001257393.1 | NP_001244322.1 | 5 | 1519 | |||
NM_001257394.1 | NP_001244323.1 | 3 | 1578 | |||
NM_001257395.1 | NP_001244324.1 | 4 | 1335 | |||
NM_001257396.1 | NP_001244325.1 | 3 | 1218 | |||
NM_001257397.1 | NP_001244326.1 | 5 | 1427 | |||
NM_001257398.1 | NP_001244327.1 | 5 | 1483 | |||
NM_001257399.1 | NP_001244328.1 | 5 | 1462 | |||
NM_001282575.2 | NP_001269504.1 | 5 | 1779 | |||
NM_001282576.1 | NP_001269505.1 | 5 | 1618 | |||
NM_001282577.2 | NP_001269506.1 | 5 | 1396 | |||
NM_001282578.2 | NP_001269507.1 | 4 | 1533 | |||
NM_001282579.1 | NP_001269508.1 | 5 | 1444 | |||
NM_021988.5 | NP_068823.2 | 4 | 1727 | |||
NM_022442.5 | NP_071887.1 | 4 | 1370 | |||
NM_199144.2 | NP_954595.1 | 5 | 1650 | |||
NR_047554.1 | 3 | 1195 | ||||
GenBank mRNA | AF049671.1 | - | 1 | 93 | ||
AL110132.1 | - | 2 | 3359 | |||
BC000468.2 | - | 4 | 1284 | 2961017 | ||
BC008944.2 | - | 3 | 1261 | |||
BX647511.1 | - | 3 | 1163 | |||
BX648147.1 | - | 3 | 1170 | |||
U39360.1 | - | 5 | 1412 | |||
U97279.1 | - | 1 | 1611 |
Gene Symbol: TMEM189-UBE2V1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199203.2 | NP_954673.1 | 8 | 2104 | ||
GenBank mRNA | BM556348.1 | - | 1 | 164 | ||
BU860198.1 | - | 1 | 466 | |||
U39361.1 | - | 5 | 1565 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
