siRNA Details
Gene Symbol: TMEM189-UBE2V1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199203.2 | NP_954673.1 | 8 | 1259 | ||
GenBank mRNA | BU156219.1 | - | 4 | 434 | ||
BU619725.1 | - | 2 | 232 | |||
CF145300.1 | - | 2 | 161 | |||
U39361.1 | - | 5 | 720 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
