siRNA Details
Gene Symbol: RPL21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000982.3 | NP_000973.2 | 1 | 378 | ||
GenBank mRNA | AF086172.1 | - | 1 | 277 | ||
AK130968.1 | - | 1 | 377 | |||
AK311795.1 | - | 1 | 375 | |||
BC001603.1 | - | 1 | 360 | 3543702 | ||
BC007505.2 | - | 1 | 341 | 2964441 | ||
BC019002.2 | - | 1 | 333 | |||
BC062981.1 | - | 1 | 358 | 6084268 | ||
BC070184.1 | - | 1 | 362 | 4281405 | ||
BC070323.1 | - | 1 | 375 | 4248655 | ||
BC070330.1 | - | 1 | 375 | 3931616 | ||
BC071902.1 | - | 1 | 358 | 4901492 | ||
BC104667.1 | - | 1 | 379 | 6724053 | ||
BX647669.1 | - | 1 | 377 | |||
CR457032.1 | - | 1 | 335 | |||
U14967.1 | - | 1 | 364 | |||
X89401.1 | - | 1 | 348 |
Gene Symbol: RPL21P19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209100.1 | - | 1 | 437 |
Gene Symbol: RPL21P28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026911.1 | 1 | 380 |
Gene Symbol: RPL21P44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027153.1 | 1 | 548 | |||
GenBank mRNA | BC057822.1 | - | 1 | 544 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
