siRNA Details
Gene Symbol: KRTAP20-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181616.1 | NP_853647.1 | 1 | 75 | ||
GenBank mRNA | AB096957.1 | - | 1 | 103 | ||
BC111084.1 | - | 1 | 75 | 40013739 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
