siRNA Details
Gene Symbol: PRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006248.3 | NP_006239.3 | 3, 4 | 1323 | ||
GenBank mRNA | K03208.1 | - | 1, 2 | 793 |
Gene Symbol: PRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005039.3 | NP_005030.2 | 3, 4 | 1070 | ||
NM_199353.2 | NP_955385.1 | 4, 5 | 671 | |||
NM_199354.2 | NP_955386.1 | 4, 5 | 611 | |||
GenBank mRNA | BC044827.1 | - | 3, 4 | 1056 | ||
BG216715.1 | - | 2, 3 | 752 | |||
K03204.1 | - | 3, 4 | 1070 | |||
K03205.1 | - | 4, 5 | 659 | |||
K03206.1 | - | 4, 5 | 607 |
Gene Symbol: PRB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261399.2 | NP_001248328.1 | 4, 5 | 596 | ||
NM_002723.5 | NP_002714.2 | 3, 4 | 803 | |||
GenBank mRNA | BC035362.1 | - | 3, 4 | 852 | ||
BC130386.1 | - | 3, 4 | 851 | 40146417 | ||
K03207.1 | - | 3, 4 | 802 |
Gene Symbol: PRH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291314.1 | NP_001278243.1 | 6, 7 | 1082 | ||
NM_001291315.1 | NP_001278244.1 | 5, 6 | 960 | |||
GenBank mRNA | AB593029.1 | - | 6, 7 | 897 | ||
BC031043.1 | - | 8, 9 | 1126 | |||
BC133676.1 | - | 4, 5 | 565 | 40119187 | ||
K03203.1 | - | 4, 5 | 570 |
Gene Symbol: PRH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC095488.1 | - | 3, 4 | 561 | 30327943 | |
BX538206.1 | - | 4, 5 | 511 | |||
BX538207.1 | - | 4, 5 | 497 | |||
BX647085.1 | - | 4, 5 | 393 | |||
BX647156.1 | - | 4, 5 | 465 | |||
BX648436.1 | - | 4, 5 | 485 | |||
BX648901.1 | - | 3, 4 | 491 | |||
CR749583.1 | - | 4, 5 | 324 | |||
K03202.1 | - | 3, 4 | 570 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
