siRNA Details
Gene Symbol: TMEM233 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136534.1 | NP_001130006.1 | 3 | 1213 | ||
GenBank mRNA | AK296289.1 | - | 3 | 1140 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
