siRNA Details
Gene Symbol: TMEM233 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136534.1 | NP_001130006.1 | 2 | 689 | ||
XM_005253880.3 | XP_005253937.1 | 2 | 2025 | |||
XM_005253881.3 | XP_005253938.1 | 2 | 2021 | |||
XM_011538328.2 | XP_011536630.1 | 2 | 2022 | |||
XM_011538329.2 | XP_011536631.1 | 2 | 2022 | |||
XM_011538330.2 | XP_011536632.1 | 2 | 2022 | |||
XM_011538331.2 | XP_011536633.1 | 2 | 2020 | |||
GenBank mRNA | AK296289.1 | - | 2 | 614 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
