siRNA Details
Gene Symbol: FAM205A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141917.1 | NP_001135389.1 | 1 | 153 | ||
GenBank mRNA | AK302320.1 | - | 1 | 83 | ||
DC402407.1 | - | 1 | 153 | |||
DC403775.1 | - | 1 | 123 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
