siRNA Details
Gene Symbol: ZNF732 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137608.1 | NP_001131080.1 | 3 | 385 | ||
GenBank mRNA | AK302099.1 | - | 3 | 385 |
Gene Symbol: ZNF141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003441.2 | NP_003432.1 | 4 | 564 | ||
XM_011513562.2 | XP_011511864.1 | 3 | 1394 | |||
XM_011513563.2 | XP_011511865.1 | 2 | 1152 | |||
XM_017008591.1 | XP_016864080.1 | 4 | 1486 | |||
GenBank mRNA | BC074853.2 | - | 4 | 424 | 30915571 | |
L15309.1 | - | 4 | 542 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
