siRNA Details
Gene Symbol: ALG1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136152.1 | NP_001129624.1 | 3, 4 | 442 | ||
GenBank mRNA | BI833803.1 | - | 3, 4 | 442 |
Gene Symbol: ALG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023457.1 | XP_016878946.1 | 7, 8 | 906 |
Gene Symbol: ALG1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015050.2 | NP_001015050.2 | 2, 3 | 272 | ||
NM_001195223.1 | NP_001182152.1 | 3, 4 | 383 | |||
XM_017005844.1 | XP_016861333.1 | 3, 4 | 3647 | |||
XM_017005845.1 | XP_016861334.1 | 3, 4 | 273 | |||
XM_017005846.1 | XP_016861335.1 | 2, 3 | 3536 | |||
XM_017005847.1 | XP_016861336.1 | 2, 3 | 162 | |||
GenBank mRNA | BC073816.1 | - | 2, 3 | 267 | 5419254 | |
BI085096.1 | - | 3, 4 | 375 | |||
CD685738.1 | - | 2, 3 | 293 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
