siRNA Details
Gene Symbol: MXRA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008529.2 | NP_001008529.1 | 5 | 1413 | ||
NM_198530.3 | NP_940932.2 | 4 | 1332 | |||
NR_130926.1 | 4 | 1031 | ||||
NR_130927.1 | 5 | 1230 | ||||
NR_130928.1 | 4 | 1149 | ||||
XM_005257383.2 | XP_005257440.1 | 4 | 1499 | |||
XM_005257384.2 | XP_005257441.1 | 3 | 1418 | |||
GenBank mRNA | AK127801.1 | - | 1 | 2893 | ||
AK128460.1 | - | 3 | 2975 | |||
AL046940.1 | - | 1 | 83 | |||
AW888225.1 | - | 5 | 1148 | |||
BC053983.1 | - | 1 | 1332 | 5470630 | ||
BC094776.1 | - | 1 | 1303 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
