siRNA Details
Gene Symbol: MXRA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198530.3 | NP_940932.2 | 3, 4 | 562 | ||
NR_130926.1 | 3, 4 | 261 | ||||
NR_130928.1 | 3, 4 | 379 | ||||
XM_005257384.2 | XP_005257441.1 | 2, 3 | 648 | |||
GenBank mRNA | AK128460.1 | - | 2, 3 | 2205 | ||
AY426672.1 | - | 632 | ||||
BC053983.1 | - | 1 | 562 | 5470630 | ||
BC094776.1 | - | 1 | 533 | |||
BQ109532.1 | - | 2, 3 | 173 | |||
CB993617.1 | - | 560 | ||||
DA362466.1 | - | 3, 4 | 261 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
