siRNA Details
Gene Symbol: DLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164271.1 | NP_001157743.1 | 14 | 4237 | ||
NM_001316668.1 | NP_001303597.1 | 14 | 4425 | |||
NM_006094.4 | NP_006085.2 | 14 | 4495 | |||
NM_182643.2 | NP_872584.2 | 18 | 5930 | |||
XM_005273374.1 | XP_005273431.1 | 18 | 5811 | |||
XM_017012951.1 | XP_016868440.1 | 18 | 5681 | |||
XM_017012952.1 | XP_016868441.1 | 14 | 4228 | |||
GenBank mRNA | AB051510.1 | - | 18 | 5816 | ||
AK023128.1 | - | 1 | 440 | |||
AL117604.1 | - | 1 | 32 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
