siRNA Details
Gene Symbol: DLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164271.1 | NP_001157743.1 | 14 | 3310 | ||
NM_001316668.1 | NP_001303597.1 | 14 | 3498 | |||
NM_006094.4 | NP_006085.2 | 14 | 3568 | |||
NM_182643.2 | NP_872584.2 | 18 | 5003 | |||
XM_005273374.1 | XP_005273431.1 | 18 | 4884 | |||
XM_017012951.1 | XP_016868440.1 | 18 | 4754 | |||
XM_017012952.1 | XP_016868441.1 | 14 | 3301 | |||
GenBank mRNA | AB051510.1 | - | 18 | 4889 | ||
AF026219.1 | - | 14 | 3521 | |||
AF035119.1 | - | 14 | 3572 | |||
AK025544.1 | - | 14 | 3497 | |||
AK025733.1 | - | 9 | 1744 | |||
AK289734.1 | - | 14 | 3491 | |||
AK291682.1 | - | 18 | 4844 | |||
AK299049.1 | - | 14 | 3310 | |||
BC054511.1 | - | 18 | 4758 | 5240161 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
