siRNA Details
Gene Symbol: ZNF577 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135590.1 | NP_001129062.1 | 7 | 1156 | ||
NM_032679.2 | NP_116068.2 | 7 | 1309 | |||
XM_006723432.3 | XP_006723495.1 | 7 | 1333 | |||
XM_006723433.2 | XP_006723496.1 | 7 | 1336 | |||
XM_011527408.2 | XP_011525710.1 | 6 | 2292 | |||
XM_017027387.1 | XP_016882876.1 | 6 | 2115 | |||
GenBank mRNA | AK289479.1 | - | 7 | 1284 | ||
AK291812.1 | - | 7 | 1156 | |||
AL832871.1 | - | 6 | 2074 | |||
BC004992.1 | - | 7 | 1277 | 2905976 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
