siRNA Details
Gene Symbol: ZNF577 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135590.1 | NP_001129062.1 | 7 | 2652 | ||
NM_032679.2 | NP_116068.2 | 7 | 2805 | |||
GenBank mRNA | BC004992.1 | - | 7 | 2773 | 2905976 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
