siRNA Details
Gene Symbol: ZNF577 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135590.1 | NP_001129062.1 | 7 | 2057 | ||
NM_032679.2 | NP_116068.2 | 7 | 2210 | |||
XM_006723432.3 | XP_006723495.1 | 7 | 2234 | |||
XM_006723433.2 | XP_006723496.1 | 7 | 2237 | |||
XM_011527408.2 | XP_011525710.1 | 6 | 3193 | |||
XM_017027387.1 | XP_016882876.1 | 6 | 3016 | |||
GenBank mRNA | BU949047.1 | - | 1 | 31 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
