siRNA Details
Gene Symbol: DEFB108B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002035.1 | NP_001002035.1 | 2 | 76 | ||
GenBank mRNA | AF529416.1 | - | 2 | 76 | ||
AY122468.1 | - | 1 | 13 | |||
BX095678.1 | - | 3 | 313 |
Gene Symbol: DEFB108P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF540980.1 | - | 2 | 76 |
Gene Symbol: LOC100133128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017008898.1 | XP_016864387.1 | 2 | 252 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
