siRNA Details
Gene Symbol: HIST1H2BN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003520.3 | NP_003511.1 | 1 | 118 | ||
GenBank mRNA | AK312228.1 | - | 1 | 227 | ||
BC101411.1 | - | 1 | 118 | 40021691 | ||
BC101412.1 | - | 1 | 118 | 40021693 | ||
BC101413.1 | - | 1 | 118 | 40021695 |
Gene Symbol: HIST1H2BL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003519.3 | NP_003510.1 | 1 | 143 | ||
GenBank mRNA | AK312114.1 | - | 1 | 182 | ||
BC093759.1 | - | 1 | 200 | |||
BC093761.1 | - | 1 | 199 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
