siRNA Details
Gene Symbol: HIST1H2BN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003520.3 | NP_003511.1 | 1 | 355 | ||
GenBank mRNA | AK312228.1 | - | 1 | 464 | ||
BC101411.1 | - | 1 | 355 | 40021691 | ||
BC101412.1 | - | 1 | 355 | 40021693 | ||
BC101413.1 | - | 1 | 355 | 40021695 |
Gene Symbol: H2BFS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC126339.1 | - | 1 | 405 |
Gene Symbol: HIST1H2BC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003526.2 | NP_003517.2 | 1 | 355 | ||
GenBank mRNA | AK312096.1 | - | 1 | 378 | ||
BC105595.1 | - | 1 | 381 | 4734445 | ||
BC106899.2 | - | 1 | 354 |
Gene Symbol: HIST1H2BD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021063.3 | NP_066407.1 | 1 | 404 | ||
NM_138720.2 | NP_619790.1 | 1 | 404 | |||
XM_005249039.3 | XP_005249096.1 | 1 | 540 | |||
GenBank mRNA | AJ223353.1 | - | 1 | 383 | ||
AK292712.1 | - | 1 | 400 | |||
BC002842.2 | - | 1 | 380 | 3659807 | ||
BC096122.3 | - | 1 | 355 | 40002354 | ||
BM153714.1 | - | 1 | 404 |
Gene Symbol: HIST1H2BF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003522.3 | NP_003513.1 | 1 | 355 | ||
GenBank mRNA | BC096120.1 | - | 1 | 355 | 40002352 | |
BC096123.1 | - | 1 | 355 | 40002357 |
Gene Symbol: HIST1H2BK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001312653.1 | NP_001299582.1 | 1 | 415 | ||
NM_080593.2 | NP_542160.1 | 1 | 415 | |||
GenBank mRNA | AJ223352.1 | - | 1 | 371 | ||
AK292062.1 | - | 1 | 412 | |||
BC000893.1 | - | 1 | 397 | 3463930 | ||
BC051872.1 | - | 1 | 395 | 6526835 | ||
BC064959.1 | - | 1 | 395 | 6093977 | ||
BC108737.1 | - | 1 | 395 | 4707078 | ||
DA873786.1 | - | 1 | 415 |
Gene Symbol: HIST1H2BL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003519.3 | NP_003510.1 | 1 | 380 | ||
GenBank mRNA | AK312114.1 | - | 1 | 419 | ||
BC093759.1 | - | 1 | 437 | |||
BC093761.1 | - | 1 | 436 |
Gene Symbol: HIST1H2BM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003521.2 | NP_003512.1 | 1 | 355 | ||
GenBank mRNA | BC066244.1 | - | 1 | 355 | 7002058 | |
BC067485.1 | - | 1 | 355 | |||
BC067486.1 | - | 1 | 355 | 7002060 | ||
BC067487.1 | - | 1 | 355 | 7002064 | ||
BC067488.1 | - | 1 | 355 | 7002065 | ||
BC067489.1 | - | 1 | 355 | 7002066 |
Gene Symbol: LOC102724334 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006723918.3 | XP_006723981.1 | 1 | 379 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
