siRNA Details
Gene Symbol: HIST1H2BN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003520.3 | NP_003511.1 | 1 | 196 | ||
GenBank mRNA | AK312228.1 | - | 1 | 305 | ||
BC101411.1 | - | 1 | 196 | 40021691 | ||
BC101412.1 | - | 1 | 196 | 40021693 | ||
BC101413.1 | - | 1 | 196 | 40021695 |
Gene Symbol: HIST1H2BO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003527.4 | NP_003518.2 | 1 | 234 | ||
GenBank mRNA | BC106720.2 | - | 1 | 217 | 40014271 | |
BX647290.1 | - | 1 | 234 |
Gene Symbol: HIST3H2BB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175055.2 | NP_778225.1 | 1 | 219 | ||
GenBank mRNA | AK091220.1 | - | 1 | 219 | ||
BC100855.1 | - | 1 | 218 | 40002412 | ||
BC100856.1 | - | 1 | 218 | 40002414 | ||
BC100857.1 | - | 1 | 218 | 40002416 | ||
BC100858.1 | - | 1 | 218 | 40002418 | ||
BC137467.1 | - | 1 | 218 | 9021467 | ||
BC137468.1 | - | 1 | 218 | 9021468 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
