siRNA Details
Gene Symbol: TNFRSF25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039664.1 | NP_001034753.1 | 2 | 216 | ||
NM_003790.2 | NP_003781.1 | 2 | 216 | |||
NM_148965.1 | NP_683866.1 | 2 | 216 | |||
NM_148966.1 | NP_683867.1 | 2 | 216 | |||
NM_148967.1 | NP_683868.1 | 2 | 216 | |||
NM_148970.1 | NP_683871.1 | 2 | 216 | |||
GenBank mRNA | AB308321.1 | - | 2 | 128 | ||
AF026070.1 | - | 2 | 196 | |||
AF026071.1 | - | 2 | 196 | |||
AY358309.1 | - | 2 | 230 | |||
BC117189.1 | - | 2 | 196 | 40125740 | ||
BC143886.1 | - | 2 | 196 | 9052402 | ||
U72763.1 | - | 2 | 128 | |||
U74611.1 | - | 2 | 216 | |||
U75380.1 | - | 2 | 119 | |||
U75381.1 | - | 2 | 119 | |||
U78029.1 | - | 2 | 128 | |||
U83597.1 | - | 2 | 112 | |||
U83598.1 | - | 2 | 112 | |||
U94501.1 | - | 2 | 128 | |||
U94502.1 | - | 2 | 128 | |||
U94503.1 | - | 2 | 128 | |||
U94504.1 | - | 2 | 128 | |||
U94505.1 | - | 2 | 128 | |||
U94506.1 | - | 2 | 128 | |||
U94507.1 | - | 2 | 128 | |||
U94508.1 | - | 2 | 128 | |||
U94509.1 | - | 2 | 128 | |||
U94510.1 | - | 2 | 128 | |||
U94512.1 | - | 2 | 120 | |||
Y09392.1 | - | 2 | 196 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
