siRNA Details
Gene Symbol: STAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282716.1 | NP_001269645.1 | 10 | 1277 | ||
NM_001282717.1 | NP_001269646.1 | 10 | 1358 | |||
NM_001282718.1 | NP_001269647.1 | 8 | 1151 | |||
NM_012447.3 | NP_036579.2 | 10 | 1358 | |||
XM_011515742.1 | XP_011514044.1 | 10 | 1325 | |||
XM_017011683.1 | XP_016867172.1 | 10 | 1205 | |||
XM_017011684.1 | XP_016867173.1 | 10 | 1325 | |||
XM_017011685.1 | XP_016867174.1 | 10 | 1438 | |||
XM_017011686.1 | XP_016867175.1 | 10 | 1205 | |||
XM_017011687.1 | XP_016867176.1 | 8 | 1031 | |||
GenBank mRNA | AJ007798.2 | - | 10 | 1278 | ||
AK292394.1 | - | 10 | 1185 | |||
AK302587.1 | - | 7 | 769 | |||
AK302693.1 | - | 8 | 974 | |||
AL833816.1 | - | 9 | 1449 | |||
BC140932.1 | - | 10 | 1117 | 9056744 | ||
BC146806.1 | - | 10 | 1183 | 8860345 |
Gene Symbol: STAG3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040583.2 | 5 | 642 | |||
GenBank mRNA | AK289466.1 | - | 6 | 611 | ||
AK291820.1 | - | 3 | 282 |
Gene Symbol: STAG3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK309007.1 | - | 5 | 473 | ||
BI753182.1 | - | 5 | 463 |
Gene Symbol: STAG3L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040582.2 | 4 | 661 | |||
GenBank mRNA | AK303592.1 | - | 4 | 322 | ||
AK316130.1 | - | 4 | 524 | |||
AY313779.1 | - | 4 | 516 | |||
BC030509.1 | - | 1 | 46 | |||
BC066937.1 | - | 4 | 394 | 4826886 | ||
BI829811.1 | - | 4 | 396 | |||
DA505391.1 | - | 4 | 533 | |||
DC395490.1 | - | 4 | 444 | |||
DC410426.1 | - | 4 | 322 |
Gene Symbol: TRIM74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012197.1 | XP_016867686.1 | 4 | 533 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
