siRNA Details
Gene Symbol: STAG3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040584.2 | 5 | 541 | |||
GenBank mRNA | AK303473.1 | - | 6 | 503 | ||
AK307228.1 | - | 6 | 620 | |||
AK309007.1 | - | 7 | 628 | |||
BC002581.2 | - | 6 | 491 | |||
BC119761.1 | - | 3 | 242 | 40119328 | ||
BG537494.1 | - | 5 | 548 | |||
BI753182.1 | - | 7 | 618 | |||
CB956482.1 | - | 5 | 520 |
Gene Symbol: STAG3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK289466.1 | - | 8 | 766 | ||
BG829469.1 | - | 5 | 559 |
Gene Symbol: STAG3L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK316130.1 | - | 6 | 679 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
