siRNA Details
Gene Symbol: STAG3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK309007.1 | - | 5 | 495 | ||
BI753182.1 | - | 5 | 485 |
Gene Symbol: STAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282716.1 | NP_001269645.1 | 10 | 1299 | ||
NM_001282717.1 | NP_001269646.1 | 10 | 1380 | |||
NM_001282718.1 | NP_001269647.1 | 8 | 1173 | |||
NM_012447.3 | NP_036579.2 | 10 | 1380 | |||
XM_011515742.1 | XP_011514044.1 | 10 | 1347 | |||
XM_017011683.1 | XP_016867172.1 | 10 | 1227 | |||
XM_017011684.1 | XP_016867173.1 | 10 | 1347 | |||
XM_017011685.1 | XP_016867174.1 | 10 | 1460 | |||
XM_017011686.1 | XP_016867175.1 | 10 | 1227 | |||
XM_017011687.1 | XP_016867176.1 | 8 | 1053 | |||
GenBank mRNA | AJ007798.2 | - | 10 | 1300 | ||
AK292394.1 | - | 10 | 1207 | |||
AK302587.1 | - | 7 | 791 | |||
AK302693.1 | - | 8 | 996 | |||
AL833816.1 | - | 9 | 1471 | |||
BC140932.1 | - | 10 | 1139 | 9056744 | ||
BC146806.1 | - | 10 | 1205 | 8860345 |
Gene Symbol: STAG3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040583.2 | 5 | 664 | |||
GenBank mRNA | AK289466.1 | - | 6 | 633 | ||
AK291820.1 | - | 3 | 304 |
Gene Symbol: STAG3L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040582.2 | 4 | 683 | |||
GenBank mRNA | AK303592.1 | - | 4 | 344 | ||
AK316130.1 | - | 4 | 546 | |||
AY313779.1 | - | 4 | 538 | |||
BC030509.1 | - | 1 | 68 | |||
BC066937.1 | - | 4 | 416 | 4826886 | ||
BG472630.1 | - | 4 | 510 | |||
BI829811.1 | - | 4 | 418 | |||
DC395490.1 | - | 4 | 466 | |||
DC410426.1 | - | 4 | 344 |
Gene Symbol: TRIM74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012197.1 | XP_016867686.1 | 4 | 555 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
