siRNA Details
Gene Symbol: TNFSF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172088.2 | NP_742085.1 | 6, 7 | 1492 | ||
GenBank mRNA | AF114013.1 | - | 6, 7 | 997 | ||
BC008042.2 | - | 6, 7 | 847 | 3837834 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
