siRNA Details
Gene Symbol: TNFSF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198622.1 | NP_001185551.1 | 5 | 1558 | ||
NM_001198623.1 | NP_001185552.1 | 5 | 1555 | |||
NM_001198624.1 | NP_001185553.1 | 6 | 863 | |||
NM_003808.3 | NP_003799.1 | 6 | 1639 | |||
NM_172087.2 | NP_742084.1 | 5 | 1591 | |||
NR_073490.2 | 4 | 1512 | ||||
GenBank mRNA | AF046888.1 | - | 6 | 1172 | ||
AF114011.1 | - | 6 | 1172 | |||
AF114012.1 | - | 5 | 1040 | |||
AF184972.1 | - | 6 | 1016 | |||
AK090698.1 | - | 5 | 1407 | |||
AK225556.1 | - | 6 | 1183 | |||
AK301221.1 | - | 5 | 1088 | |||
AY081050.1 | - | 6 | 1163 | |||
AY358880.1 | - | 6 | 1004 | |||
DQ149579.1 | - | 5 | 840 | |||
EF211088.1 | - | 4 | 794 |
Gene Symbol: TNFSF12-TNFSF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172089.3 | NP_742086.1 | 11 | 1227 | ||
GenBank mRNA | AY081051.1 | - | 11 | 1186 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
