siRNA Details
Gene Symbol: INAFM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178511.5 | NP_848606.3 | 1 | 151 | ||
GenBank mRNA | BC022076.2 | - | 1 | 151 | ||
BC045749.1 | - | 1 | 151 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
