siRNA Details
Gene Symbol: CLEC18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011880.2 | NP_001011880.2 | 8, 9 | 1180 | ||
XM_005255976.2 | XP_005256033.1 | 8, 9 | 1257 | |||
XM_011523093.2 | XP_011521395.1 | 7, 8 | 963 | |||
XM_017023255.1 | XP_016878744.1 | 9, 10 | 1434 | |||
GenBank mRNA | AY358373.1 | - | 8, 9 | 1102 | ||
BM695392.1 | - | 4, 5 | 391 |
Gene Symbol: CLEC18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136214.2 | NP_001129686.1 | 9, 10 | 1118 | ||
NM_001271197.1 | NP_001258126.1 | 9, 10 | 1198 | |||
NM_182619.3 | NP_872425.2 | 9, 10 | 1224 | |||
XM_005255934.1 | XP_005255991.1 | 8, 9 | 1167 | |||
XM_017023206.1 | XP_016878695.1 | 8, 9 | 1167 | |||
XM_017023207.1 | XP_016878696.1 | 7, 8 | 997 | |||
XM_017023208.1 | XP_016878697.1 | 8, 9 | 1350 | |||
GenBank mRNA | AF521893.1 | - | 9, 10 | 1237 | ||
AK074773.1 | - | 8, 9 | 1052 | |||
AK289884.1 | - | 8, 9 | 1166 | |||
BC078143.1 | - | 9, 10 | 1100 | 5761001 | ||
BC141808.1 | - | 9, 10 | 1198 | 40147524 |
Gene Symbol: CLEC18C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173619.3 | NP_775890.2 | 8, 9 | 1086 | ||
XM_006721179.1 | XP_006721242.1 | 8, 9 | 1182 | |||
XM_011523034.1 | XP_011521336.1 | 9, 10 | 1428 | |||
XM_011523036.2 | XP_011521338.1 | 7, 8 | 1012 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
