siRNA Details
Gene Symbol: IGSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170961.1 | NP_001164432.1 | 14 | 2724 | ||
NM_001170962.1 | NP_001164433.1 | 13 | 2682 | |||
NM_001555.4 | NP_001546.2 | 14 | 2709 | |||
XM_011531330.1 | XP_011529632.1 | 15 | 2622 | |||
XM_011531333.1 | XP_011529635.1 | 14 | 2724 | |||
XM_011531334.1 | XP_011529636.1 | 14 | 2724 | |||
GenBank mRNA | AF034198.1 | - | 13 | 2531 | ||
AK226008.1 | - | 14 | 2581 | |||
AK295438.1 | - | 5 | 760 | |||
AK307871.1 | - | 14 | 2585 | |||
BC063884.1 | - | 14 | 2545 | 30387876 | ||
Y10523.1 | - | 13 | 2506 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
