siRNA Details
Gene Symbol: IGSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170963.1 | NP_001164434.1 | 5 | 1731 | ||
NM_205833.3 | NP_991402.1 | 5 | 1907 | |||
GenBank mRNA | AB058894.2 | - | 5 | 1760 | ||
AK057916.1 | - | 5 | 1738 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
