siRNA Details
Gene Symbol: ARL17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016632.2 | NP_057716.2 | 5 | 561 | ||
XM_011524875.2 | XP_011523177.1 | 5 | 960 | |||
GenBank mRNA | AF208864.1 | - | 5 | 510 |
Gene Symbol: ARL17B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011524160.2 | XP_011522462.1 | 5 | 978 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
