siRNA Details
Gene Symbol: PCDHGC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002588.3 | NP_002579.2 | 1 | 501 | ||
NM_032402.1 | NP_115778.1 | 1 | 501 | |||
GenBank mRNA | AF152337.1 | - | 1 | 386 | ||
AF152524.1 | - | 1 | 386 | |||
BC019299.1 | - | 1 | 500 | 2905509 | ||
BC026218.1 | - | 1 | 490 | 4153558 | ||
L11373.1 | - | 1 | 500 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
