siRNA Details
Gene Symbol: KRTAP10-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198689.2 | NP_941962.1 | 1 | 444 | ||
GenBank mRNA | AB076354.1 | - | 1 | 444 | ||
BC119643.2 | - | 1 | 441 | 40115724 |
Gene Symbol: KRTAP10-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198687.2 | NP_941960.2 | 1 | 449 | ||
GenBank mRNA | AB076351.1 | - | 1 | 449 | ||
BC125048.1 | - | 1 | 438 | 40116133 | ||
BC125049.1 | - | 1 | 438 | 40116140 |
Gene Symbol: KRTAP10-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198688.3 | NP_941961.3 | 1 | 440 | ||
GenBank mRNA | AB076353.1 | - | 1 | 440 | ||
BC140725.1 | - | 453 | 9021593 |
Gene Symbol: KRTAP10-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198695.2 | NP_941968.2 | 1 | 471 | ||
GenBank mRNA | AB076355.1 | - | 1 | 471 | ||
BC127016.1 | - | 1 | 465 | 40119338 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
