siRNA Details
Gene Symbol: TRIM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033342.3 | NP_203128.1 | 2 | 839 | ||
NM_203295.1 | NP_976040.1 | 2 | 453 | |||
XM_017009903.1 | XP_016865392.1 | 2 | 810 | |||
XM_017009904.1 | XP_016865393.1 | 2 | 1546 | |||
GenBank mRNA | AF220032.1 | - | 2 | 839 | ||
AF396653.1 | - | 2 | 453 | |||
AK314872.1 | - | 2 | 702 | |||
BC011567.1 | - | 2 | 692 | |||
BC080553.1 | - | 2 | 669 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
