siRNA Details
Gene Symbol: PRSS41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135086.1 | NP_001128558.1 | 3 | 380 | ||
XM_011522477.2 | XP_011520779.1 | 5 | 998 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
