siRNA Details
Gene Symbol: POTEH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136213.1 | NP_001129685.1 | 5 | 1136 | ||
GenBank mRNA | AY466021.1 | - | 6 | 1335 |
Gene Symbol: POTEE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083538.1 | NP_001077007.1 | 5 | 1025 | ||
XM_011511216.2 | XP_011509518.1 | 5 | 1103 | |||
XM_017004159.1 | XP_016859648.1 | 5 | 1134 | |||
XM_017004161.1 | XP_016859650.1 | 7 | 1462 | |||
GenBank mRNA | AY462868.1 | - | 5 | 1025 | ||
AY462871.1 | - | 5 | 1025 | |||
AY462872.1 | - | 5 | 1055 | |||
AY462873.1 | - | 5 | 1025 |
Gene Symbol: POTEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099771.2 | NP_001093241.1 | 7 | 1373 | ||
XM_017004833.1 | XP_016860322.1 | 5 | 1148 | |||
GenBank mRNA | EF523384.1 | - | 7 | 1375 |
Gene Symbol: POTEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005356.2 | NP_001005356.1 | 5 | 1025 | ||
NR_027480.1 | 5 | 1025 | ||||
GenBank mRNA | AY462874.1 | - | 5 | 1025 | ||
AY465171.1 | - | 7 | 1283 | |||
AY465172.1 | - | 6 | 1025 | |||
BC127623.1 | - | 5 | 1005 |
Gene Symbol: POTEI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277406.1 | NP_001264335.1 | 5 | 973 | ||
XM_011511670.2 | XP_011509972.1 | 5 | 1145 | |||
XM_017004732.1 | XP_016860221.1 | 6 | 1124 |
Gene Symbol: POTEJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277083.1 | NP_001264012.1 | 5 | 862 | ||
XM_011511683.2 | XP_011509985.1 | 5 | 1034 | |||
XM_017004741.1 | XP_016860230.1 | 6 | 1007 |
Gene Symbol: POTEM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145442.1 | NP_001138914.1 | 5 | 1025 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
