siRNA Details
Gene Symbol: SPAG11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016512.3 | NP_057596.1 | 2 | 344 | ||
NM_058200.2 | NP_478107.2 | 2 | 344 | |||
NM_058201.2 | NP_478108.2 | 2 | 344 | |||
NM_058202.2 | NP_478109.1 | 2 | 344 | |||
NM_058203.2 | NP_478110.1 | 2 | 344 | |||
GenBank mRNA | AF168616.1 | - | 2 | 344 | ||
AF168617.1 | - | 2 | 328 | |||
AJ628017.1 | - | 2 | 182 | |||
AJ635328.1 | - | 2 | 334 | |||
AL133652.1 | - | 2740 |
Gene Symbol: SPAG11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081552.2 | NP_001075021.2 | 2 | 344 | ||
XM_005272391.4 | XP_005272448.2 | 2 | 317 | |||
XM_005272392.4 | XP_005272449.2 | 2 | 317 | |||
XM_005272393.4 | XP_005272450.2 | 2 | 317 | |||
GenBank mRNA | AF168618.1 | - | 2 | 200 | ||
AF168619.1 | - | 2 | 200 | |||
AF168620.1 | - | 1 | 114 | |||
AF170797.1 | - | 1 | 80 | |||
BC125141.1 | - | 2 | 292 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
