siRNA Details
Gene Symbol: SPAG11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016512.3 | NP_057596.1 | 4 | 745 | ||
NM_058201.2 | NP_478108.2 | 3 | 669 | |||
NM_058206.3 | NP_478113.2 | 3 | 471 | |||
NM_058207.2 | NP_478114.2 | 2 | 395 |
Gene Symbol: SPAG11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC058833.1 | - | 4 | 621 | 6615920 | |
BC125141.1 | - | 4 | 693 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
